Assay Details
| Gene Symbol: TEKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119667.2 | XP_013975142.1 | 8 - 9 | 1380 | 62 | |
| XM_014119668.2 | XP_013975143.1 | 8 - 9 | 1116 | 62 | ||
| XM_022427847.1 | XP_022283555.1 | 8 - 9 | 1401 | 62 | ||
| XM_022427848.1 | XP_022283556.1 | 7 - 8 | 1286 | 62 | ||
| XM_539596.7 | XP_539596.4 | 8 - 9 | 1134 | 62 | ||
More Information
Set Membership: |
|

TEKT2
Genomic Map