Assay Details
| Gene Symbol: IQCA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119790.2 | XP_013975265.1 | 782 | 73 | ||
| XM_022403872.1 | XP_022259580.1 | 783 | 73 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: IQCA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014119790.2 | XP_013975265.1 | 782 | 73 | ||
| XM_022403872.1 | XP_022259580.1 | 783 | 73 | |||
Set Membership: |
|