Assay Details
| Gene Symbol: SYTL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432546.3 | XP_003432594.1 | 2991 | 99 | ||
| XM_005615547.2 | XP_005615604.1 | 3005 | 99 | |||
| XM_005615548.2 | XP_005615605.1 | 2982 | 99 | |||
| XM_005615549.2 | XP_005615606.1 | 2885 | 99 | |||
| XM_022418452.1 | XP_022274160.1 | 2862 | 99 | |||
| XM_022418462.1 | XP_022274170.1 | 3003 | 99 | |||
More Information
Set Membership: |
|

SYTL3
Genomic Map