Assay Details
| Gene Symbol: BVES | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627668.3 | XP_005627725.1 | 6 - 7 | 939 | 70 | |
| XM_005627669.3 | XP_005627726.1 | 6 - 7 | 893 | 70 | ||
| XM_022426410.1 | XP_022282118.1 | 6 - 7 | 852 | 70 | ||
| XM_539065.6 | XP_539065.3 | 6 - 7 | 910 | 70 | ||
More Information
Set Membership: |
|

BVES
Genomic Map