Assay Details
| Gene Symbol: MOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615415.2 | XP_005615472.1 | 995 | 79 | ||
| XM_541103.5 | XP_541103.2 | 993 | 79 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MOXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615415.2 | XP_005615472.1 | 995 | 79 | ||
| XM_541103.5 | XP_541103.2 | 993 | 79 | |||
Set Membership: |
|