Assay Details
| Gene Symbol: CYP2S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425478.1 | XP_022281186.1 | 8 - 9 | 1647 | 97 | |
| XM_541605.4 | XP_541605.2 | 8 - 9 | 1661 | 97 | ||
More Information
Set Membership: |
|

CYP2S1
Genomic Map