Assay Details
| Gene Symbol: SLC9A3R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001314137.1 | NP_001301066.1 | 810 | 105 | ||
| XM_014116307.2 | XP_013971782.1 | 1023 | 105 | |||
| GenBank mRNA | AB679832.1 | - | 810 | 105 | ||
More Information
Set Membership: |
|

SLC9A3R1
Genomic Map