Assay Details
| Gene Symbol: SQSTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117639.2 | XP_013973114.1 | 330 | 76 | ||
| XM_022425090.1 | XP_022280798.1 | 330 | 76 | |||
| XM_022425091.1 | XP_022280799.1 | 739 | 76 | |||
More Information
Set Membership: |
|

SQSTM1
Genomic Map