Assay Details
| Gene Symbol: NEU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627165.3 | XP_005627222.1 | 4 - 5 | 601 | 71 | |
| XM_538838.6 | XP_538838.2 | 3 - 4 | 702 | 71 | ||
| GenBank mRNA | EU179543.1 | - | 1 - 2 | 65 | 71 | |
More Information
Set Membership: |
|

NEU1
Genomic Map