Assay Details
| Gene Symbol: ADAMTSL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407276.1 | XP_022262984.1 | 8 - 9 | 1463 | 61 | |
| XM_022407278.1 | XP_022262986.1 | 8 - 9 | 1465 | 61 | ||
More Information
Set Membership: |
|

ADAMTSL5
Genomic Map