Assay Details
| Gene Symbol: ZSCAN18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616064.3 | XP_005616121.1 | 6 - 7 | 1114 | 102 | |
| XM_005616065.3 | XP_005616122.1 | 6 - 7 | 1056 | 102 | ||
| XM_005616066.3 | XP_005616123.1 | 6 - 7 | 1007 | 102 | ||
More Information
Set Membership: |
|

ZSCAN18
Genomic Map