Assay Details
| Gene Symbol: TRMT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014106915.2 | XP_013962390.1 | 6 - 7 | 687 | 67 | |
| XM_014106916.2 | XP_013962391.1 | 6 - 7 | 676 | 67 | ||
| XM_022409215.1 | XP_022264923.1 | 5 - 6 | 572 | 67 | ||
| XM_022409216.1 | XP_022264924.1 | 6 - 7 | 657 | 67 | ||
More Information
Set Membership: |
|

TRMT6
Genomic Map