Assay Details
| Gene Symbol: SETD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410697.1 | XP_022266405.1 | 4 - 5 | 540 | 63 | |
| XM_022410701.1 | XP_022266409.1 | 4 - 5 | 452 | 63 | ||
More Information
Set Membership: |
|

SETD6
Genomic Map