Assay Details
| Gene Symbol: CTSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410039.1 | XP_022265747.1 | 2 - 3 | 161 | 91 | |
| XM_543203.6 | XP_543203.3 | 5 - 6 | 578 | 91 | ||
More Information
Set Membership: |
|

CTSB
Genomic Map