Assay Details
| Gene Symbol: ESCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409840.1 | XP_022265548.1 | 5 - 6 | 1168 | 98 | |
| XM_543224.6 | XP_543224.2 | 5 - 6 | 1158 | 98 | ||
More Information
Set Membership: |
|

ESCO2
Genomic Map