Assay Details
| Gene Symbol: LOC487083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617058.2 | XP_005617115.1 | 469 | 107 | ||
| XM_022407134.1 | XP_022262842.1 | 469 | 107 | |||
More Information
Set Membership: |
|

LOC487083
Genomic Map