Assay Details
| Gene Symbol: E2F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634944.3 | XP_005635001.1 | 2 - 3 | 390 | 120 | |
| XM_005634945.2 | XP_005635002.1 | 2 - 3 | 390 | 120 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

E2F1
Genomic Map