Assay Details
| Gene Symbol: TAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407989.1 | XP_022263697.1 | 2673 | 63 | ||
| XM_544263.6 | XP_544263.2 | 2876 | 63 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TAF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407989.1 | XP_022263697.1 | 2673 | 63 | ||
| XM_544263.6 | XP_544263.2 | 2876 | 63 | |||
Set Membership: |
|