Assay Details
| Gene Symbol: GNB1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410827.1 | XP_022266535.1 | 3 - 4 | 393 | 54 | |
| XM_543544.6 | XP_543544.4 | 4 - 5 | 471 | 54 | ||
More Information
Set Membership: |
|

GNB1L
Genomic Map