Assay Details
| Gene Symbol: OVOL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022409220.1 | XP_022264928.1 | 3 - 4 | 873 | 147 | |
| XM_022409221.1 | XP_022264929.1 | 2 - 3 | 744 | 147 | ||
More Information
Set Membership: |
|

OVOL2
Genomic Map