Assay Details
| Gene Symbol: AQP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022412733.1 | XP_022268441.1 | 3 - 4 | 327 | 63 | |
| XM_544701.5 | XP_544701.1 | 3 - 4 | 745 | 63 | ||
More Information
Set Membership: |
|

AQP9
Genomic Map