Assay Details
| Gene Symbol: CRYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107417.2 | XP_013962892.1 | 702 | 61 | ||
| XM_022409683.1 | XP_022265391.1 | 549 | 61 | |||
| XM_543175.6 | XP_543175.2 | 555 | 61 | |||
More Information
Set Membership: |
|

CRYL1
Genomic Map