Assay Details
| Gene Symbol: RPL22L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_545287.5 | XP_545287.2 | 2 - 3 | 177 | 111 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL22L1| Gene Symbol: RPL22L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_545287.5 | XP_545287.2 | 2 - 3 | 177 | 111 | |
Set Membership: |
|