Assay Details
| Gene Symbol: CHRNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418516.1 | XP_022274224.1 | 3 - 4 | 282 | 61 | |
| XM_546591.4 | XP_546591.2 | 3 - 4 | 321 | 61 | ||
More Information
Set Membership: |
|

CHRNB1
Genomic Map