Assay Details
| Gene Symbol: TUBA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022410870.1 | XP_022266578.1 | 2 - 3 | 317 | 69 | |
| XM_022410871.1 | XP_022266579.1 | 4 - 5 | 635 | 69 | ||
| XM_543889.5 | XP_543889.2 | 3 - 4 | 511 | 69 | ||
More Information
Set Membership: |
|

TUBA8
Genomic Map