Assay Details
| Gene Symbol: KCNMB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414142.1 | XP_022269850.1 | 2 - 3 | 298 | 74 | |
| XM_545302.5 | XP_545302.3 | 2 - 3 | 321 | 74 | ||
More Information
Set Membership: |
|

KCNMB2
Genomic Map