Assay Details
| Gene Symbol: AADAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635634.3 | XP_005635691.3 | 1121 | 105 | ||
| XM_543185.6 | XP_543185.6 | 1186 | 105 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: AADAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635634.3 | XP_005635691.3 | 1121 | 105 | ||
| XM_543185.6 | XP_543185.6 | 1186 | 105 | |||
Set Membership: |
|