Assay Details
| Gene Symbol: CCDC122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022408327.1 | XP_022264035.1 | 5 - 6 | 779 | 98 | |
| XM_022408328.1 | XP_022264036.1 | 5 - 6 | 762 | 98 | ||
| XM_022408329.1 | XP_022264037.1 | 5 - 6 | 881 | 98 | ||
| XM_022408331.1 | XP_022264039.1 | 5 - 6 | 740 | 98 | ||
| XM_022408332.1 | XP_022264040.1 | 6 - 7 | 855 | 98 | ||
| XM_022408333.1 | XP_022264041.1 | 6 - 7 | 1008 | 98 | ||
| XM_022408335.1 | XP_022264043.1 | 4 - 5 | 744 | 98 | ||
| XM_542579.6 | XP_542579.4 | 6 - 7 | 1043 | 98 | ||
More Information
Set Membership: |
|

CCDC122
Genomic Map