Assay Details
| Gene Symbol: CA5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003640227.3 | XP_003640275.1 | 4 - 5 | 654 | 88 | |
| XM_022416368.1 | XP_022272076.1 | 8 - 9 | 1582 | 88 | ||
| XM_022416369.1 | XP_022272077.1 | 8 - 9 | 1233 | 88 | ||
| XM_022416370.1 | XP_022272078.1 | 5 - 6 | 719 | 88 | ||
| XM_022416371.1 | XP_022272079.1 | 5 - 6 | 626 | 88 | ||
| XM_022416372.1 | XP_022272080.1 | 7 - 8 | 1386 | 88 | ||
More Information
Set Membership: |
|

CA5B
Genomic Map