Assay Details
Gene Symbol: WDR53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005639600.2 | XP_005639657.1 | 2 - 3 | 596 | 68 | |
XM_005639601.2 | XP_005639658.1 | 2 - 3 | 719 | 68 | ||
XM_005639602.3 | XP_005639659.1 | 2 - 3 | 634 | 68 | ||
XM_014110278.2 | XP_013965753.1 | 2 - 3 | 640 | 68 | ||
XM_545153.6 | XP_545153.1 | 2 - 3 | 630 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
