Assay Details
| Gene Symbol: LOC489992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420039.1 | XP_022275747.1 | 17 - 18 | 1917 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC489992| Gene Symbol: LOC489992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022420039.1 | XP_022275747.1 | 17 - 18 | 1917 | 62 | |
Set Membership: |
|