Assay Details
| Gene Symbol: DNTTIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_547275.6 | XP_547275.3 | 4 - 5 | 2036 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNTTIP2| Gene Symbol: DNTTIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_547275.6 | XP_547275.3 | 4 - 5 | 2036 | 76 | |
Set Membership: |
|