Assay Details
| Gene Symbol: PHPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625080.3 | XP_005625137.1 | 2 - 3 | 348 | 72 | |
| XM_022423551.1 | XP_022279259.1 | 2 - 3 | 393 | 72 | ||
More Information
Set Membership: |
|

PHPT1
Genomic Map