Assay Details
| Gene Symbol: ZNF706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014118551.2 | XP_013974026.1 | 395 | 83 | ||
| XM_022426589.1 | XP_022282297.1 | 163 | 83 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ZNF706 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014118551.2 | XP_013974026.1 | 395 | 83 | ||
| XM_022426589.1 | XP_022282297.1 | 163 | 83 | |||
Set Membership: |
|