Assay Details
| Gene Symbol: LOC489534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418646.1 | XP_022274354.1 | 1 - 2 | 214 | 68 | |
| XM_546653.6 | XP_546653.2 | 1 - 2 | 202 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022418646.1 transcripts.
Set Membership: |
|

LOC489534
Genomic Map