Assay Details
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313876.1 | NP_001300805.1 | 198 | 64 | ||
| XM_022422593.1 | XP_022278301.1 | 288 | 64 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PPY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313876.1 | NP_001300805.1 | 198 | 64 | ||
| XM_022422593.1 | XP_022278301.1 | 288 | 64 | |||
Set Membership: |
|