Assay Details
| Gene Symbol: RHBDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419579.1 | XP_022275287.1 | 836 | 72 | ||
| XM_022419580.1 | XP_022275288.1 | 677 | 72 | |||
| XM_546933.5 | XP_546933.3 | 655 | 72 | |||
More Information
Set Membership: |
|

RHBDD2
Genomic Map