Assay Details
| Gene Symbol: DAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414350.1 | XP_022270058.1 | 290 | 60 | ||
| XM_843925.5 | XP_849018.1 | 1 | 256 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414350.1 | XP_022270058.1 | 290 | 60 | ||
| XM_843925.5 | XP_849018.1 | 1 | 256 | 60 | ||
Set Membership: |
|