Assay Details
| Gene Symbol: EFCAB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624205.2 | XP_005624262.1 | 1156 | 110 | ||
| XM_005624209.2 | XP_005624266.1 | 1102 | 110 | |||
| XM_005624211.2 | XP_005624268.1 | 1084 | 110 | |||
| XM_005624213.3 | XP_005624270.1 | 790 | 110 | |||
| XM_005624214.2 | XP_005624271.1 | 874 | 110 | |||
| XM_005624215.3 | XP_005624272.1 | 432 | 110 | |||
| XM_022422783.1 | XP_022278491.1 | 790 | 110 | |||
| XM_022422784.1 | XP_022278492.1 | 771 | 110 | |||
| XM_022422785.1 | XP_022278493.1 | 735 | 110 | |||
| XM_022422786.1 | XP_022278494.1 | 1044 | 110 | |||
| XM_022422787.1 | XP_022278495.1 | 1102 | 110 | |||
| XM_022422788.1 | XP_022278496.1 | 1138 | 110 | |||
| XM_022422789.1 | XP_022278497.1 | 1048 | 110 | |||
| XM_548039.6 | XP_548039.2 | 626 | 110 | |||
More Information
Set Membership: |
|

EFCAB3
Genomic Map