Assay Details
| Gene Symbol: PIPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624876.3 | XP_005624933.1 | 2 - 3 | 343 | 65 | |
| XM_022423355.1 | XP_022279063.1 | 2 - 3 | 342 | 65 | ||
| XM_022423356.1 | XP_022279064.1 | 2 - 3 | 341 | 65 | ||
| XM_548297.6 | XP_548297.2 | 2 - 3 | 341 | 65 | ||
More Information
Set Membership: |
|

PIPOX
Genomic Map