Assay Details
| Gene Symbol: CALY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637908.2 | XP_005637965.1 | 4 - 5 | 523 | 73 | |
| XM_005637909.2 | XP_005637966.1 | 4 - 5 | 583 | 73 | ||
| XM_022411755.1 | XP_022267463.1 | 5 - 6 | 524 | 73 | ||
More Information
Set Membership: |
|

CALY
Genomic Map