Assay Details
| Gene Symbol: SH3BGRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641530.3 | XP_005641587.1 | 2 - 3 | 442 | 131 | |
| XM_005641531.3 | XP_005641588.1 | 2 - 3 | 286 | 131 | ||
| XM_022415749.1 | XP_022271457.1 | 2 - 3 | 253 | 131 | ||
More Information
Set Membership: |
|

SH3BGRL
Genomic Map