Assay Details
| Gene Symbol: TRIM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418606.1 | XP_022274314.1 | 4 - 5 | 847 | 74 | |
| XM_546637.5 | XP_546637.2 | 4 - 5 | 1068 | 74 | ||
More Information
Set Membership: |
|

TRIM16
Genomic Map