Assay Details
| Gene Symbol: MMP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620395.3 | XP_005620452.1 | 1508 | 79 | ||
| XM_843797.5 | XP_848890.1 | 1494 | 79 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: MMP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620395.3 | XP_005620452.1 | 1508 | 79 | ||
| XM_843797.5 | XP_848890.1 | 1494 | 79 | |||
Set Membership: |
|