Assay Details
| Gene Symbol: RPP21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252381.1 | NP_001239310.1 | 3 - 4 | 251 | 127 | |
| XM_005627042.3 | XP_005627099.2 | 3 - 4 | 320 | 127 | ||
| GenBank mRNA | DN370541.1 | - | 3 - 4 | 251 | 127 | |
| DN378554.1 | - | 3 - 4 | 226 | 127 | ||
More Information
Set Membership: |
|

RPP21
Genomic Map