Assay Details
| Gene Symbol: SCML1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111799.2 | XP_013967274.2 | 1 - 2 | 272 | 58 | |
| XM_014111803.2 | XP_013967278.1 | 1 - 2 | 270 | 58 | ||
| XM_022415761.1 | XP_022271469.1 | 1 - 2 | 267 | 58 | ||
| XM_022415762.1 | XP_022271470.1 | 1 - 2 | 268 | 58 | ||
| XM_022415763.1 | XP_022271471.1 | 1 - 2 | 269 | 58 | ||
| XM_022415764.1 | XP_022271472.1 | 1 - 2 | 270 | 58 | ||
| XM_022415765.1 | XP_022271473.1 | 1 - 2 | 266 | 58 | ||
| XM_022415766.1 | XP_022271474.1 | 1 - 2 | 267 | 58 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014111799.2, XM_014111803.2, XM_022415761.1, XM_022415762.1, XM_022415763.1, XM_022415764.1, XM_022415765.1, XM_022415766.1 transcripts.
Set Membership: |
|

SCML1
Genomic Map