Assay Details
Gene Symbol: ZNF248 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618775.3 | XP_005618832.1 | 451 | 84 | ||
XM_005618778.3 | XP_005618835.1 | 389 | 84 | |||
XM_022417365.1 | XP_022273073.1 | 462 | 84 | |||
XM_022417366.1 | XP_022273074.1 | 1176 | 84 | |||
XM_022417367.1 | XP_022273075.1 | 1611 | 84 | |||
XM_022417368.1 | XP_022273076.1 | 1004 | 84 | |||
XM_022417369.1 | XP_022273077.1 | 800 | 84 | |||
XM_843150.5 | XP_848243.1 | 902 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
