Assay Details
| Gene Symbol: LHX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022423854.1 | XP_022279562.1 | 2 - 3 | 300 | 102 | |
| XM_022423855.1 | XP_022279563.1 | 2 - 3 | 466 | 102 | ||
| XM_858575.4 | XP_863668.2 | 2 - 3 | 601 | 102 | ||
More Information
Set Membership: |
|

LHX2
Genomic Map