Assay Details
| Gene Symbol: TNFRSF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190744.1 | NP_001177673.1 | 366 | 61 | ||
| XM_005620342.3 | XP_005620399.1 | 394 | 61 | |||
| XM_022418057.1 | XP_022273765.1 | 280 | 61 | |||
| GenBank mRNA | FJ969445.1 | - | 366 | 61 | ||
More Information
Set Membership: |
|

TNFRSF18
Genomic Map