Assay Details
| Gene Symbol: S1PR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_547259.6 | XP_547259.2 | 1 - 2 | 312 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_547259.6 transcripts.
Set Membership: |
|

S1PR1
Genomic Map